Learn More
Adooq Bioscience LLC ELLAGIC ACID 500 MG
Catalog No. NC1906356
Encompass_Preferred
CAS NO. 476-66-4. llagic acid is a natural phenol antioxidant found in numerous fruits and vegetables. The antiproliferative and antioxidant properties of ellagic acid have spurred preliminary research into the potential health benefits of ellagic acid consumption.
Catalog No. NC1906356 Supplier Adooq Bioscience LLC Supplier No. A10344
CAS NO. 476-66-4. llagic acid is a natural phenol antioxidant found in numerous fruits and vegetables. The antiproliferative and antioxidant properties of ellagic acid have spurred preliminary research into the potential health benefits of ellagic acid consumption.