Learn More
Medchemexpress LLC HY-B0183 500mg Medchemexpress, Ellagic acid CAS:476-66-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0183500MG
Encompass_Preferred
Medchemexpress, HY-B0183 500mg Ellagic acid CAS:476-66-4 Ellagic acid is a natural antioxidant, and acts as a potent and ATP-competitive CK2 inhibitor, with an IC50 of 40 nM and a Ki of 20 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2400