Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ellagic acid, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014247 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4247 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4247 Supplier MilliporeSigma Supelco Supplier No. 1466850MG
Add to Cart
Add to Cart

Ellagic acid is a dimeric derivative of gallic acid. It is a plant polyphenol which has properties like antioxident, anti-inflammatory, anticancer and antiatherosclerosis.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 476-66-4
Physical Form Neat
Linear Formula C14H6O8
MDL Number MFCD00006914
Synonym 4,4 ′,5,5 ′,6,6 ′-Hexahydroxydiphenic acid 2,6,2 ′,6 ′-dilactone; Alizarin Yellow; Benzoaric acid; Elagostasine; Eleagic acid; Gallogen; Lagistase
Type Analytical Standard
Quantity 50 mg
Formula Weight 302.19
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.