Learn More
Abcam Ellagic acid, casein kinase 2 (CK2) inhibitor, 50MG
Catalog No. 502707412
Encompass_Preferred
MW 302.2 g/mol, Purity >99%. Selective, ATP-competitive casein kinase 2 (CK2) inhibitor (Ki = 20 nM). Exhibits a multitude of biological activities including antioxidant, antiproliferative and pro-apoptotic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7412 Supplier Abcam Supplier No. AB12062250MG
MW 302.2 g/mol, Purity >99%. Selective, ATP-competitive casein kinase 2 (CK2) inhibitor (Ki = 20 nM). Exhibits a multitude of biological activities including antioxidant, antiproliferative and pro-apoptotic activity.
The product is subject to the following: Abcam Restricted Use Statement