Learn More
Cayman Chemical Ellagic Acid, Purity: ≥95%, CAS: 476-66-4, 1 g
SDP
Catalog No. 502889179
Encompass_Preferred
A polyphenolic antioxidant that is abundant in many fruits, vegetables, plant bark, and peels; has anti-carcinogenic, anti-mutagenic, anti-inflammatory, and organ-preserving properties; blocks methylation of H3R17 by CARM1 without significantly altering histone acetylase or DNA methyltransferase activity
Catalog No. 50-288-9179 Supplier Cayman Chemical Supplier No. 105691 G
A polyphenolic antioxidant that is abundant in many fruits, vegetables, plant bark, and peels; has anti-carcinogenic, anti-mutagenic, anti-inflammatory, and organ-preserving properties; blocks methylation of H3R17 by CARM1 without significantly altering histone acetylase or DNA methyltransferase activity