Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ellagic Acid, MP Biomedicals™

Catalog No. MP215102905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215102905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215102905 Supplier MP Biomedicals, Inc Supplier No. 0215102905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the assay of factor XIIa in plasma. Ellagic Acid, MP Biomedicals is a commonly occurring plant polyphenol and an inhibitor of glutathione S-transferase with contact activation in blood coagulation.

Ellagic acid is a naturally occurring plant phenol that is a potent antioxidant, presumably by acting as a scavenger of oxygen species produced by hydrogen peroxide treatment. It acts as a protector of the DNA double helix from alkylating agent injury.

  • Delayed cutaneous hypersensitivity reaction to tuberculin and bovine serum albumin in guinea pigs
  • Has anti-mutagenic and anti-carcinogenic properties
  • Reported to inhibit topoisomerases I and II and gyrase
  • Inhibitor of glutathione S-transferase
  • Hemostatic agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Melting Point >300°C (literature)
Quantity 5 g
Solubility Information Soluble in 1M Sodium Hydroxide (10mg/mL, dark brown to green solution) or other alkalies; soluble in pyridine. Slightly soluble in water or ethanol.
Formula Weight 302.2
Physical Form Powder
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.