Learn More
Chemglass Life Sciences Ellipso-Spoon, 15 x 35 x 150mm
SDP
Catalog No. 50872450
Encompass_Preferred
Spoon size/15 x 35, Overall Length/150mm, Minimum Joint Clearance/24/40. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 5mm diameter handle.
Catalog No. 50-872-450 Supplier Chemglass Life Sciences Supplier No. CG198701
Spoon size/15 x 35, Overall Length/150mm, Minimum Joint Clearance/24/40. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 5mm diameter handle.