Learn More
Chemglass Life Sciences Ellipso-Spoon, 28 x 65 x 300mm
SDP
Catalog No. 50872452
Encompass_Preferred
Ellipso-Spoon, 28 x 65 x 300mm. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 8mm diameter handle.
Catalog No. 50-872-452 Supplier Chemglass Life Sciences Supplier No. CG198703
Ellipso-Spoon, 28 x 65 x 300mm. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 8mm diameter handle.