Learn More
Chemglass Life Sciences Ellipso-Spoon, 28 x 65 x 400mm
SDP
Catalog No. 50872453
Encompass_Preferred
Ellipso-Spoon, 28 x 65 x 400mm. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 8mm diameter handle.
Catalog No. 50-872-453 Supplier Chemglass Life Sciences Supplier No. CG198704
Ellipso-Spoon, 28 x 65 x 400mm. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 8mm diameter handle.