Learn More
Electron Microscopy Sciences Ellipso-Spoon Sampler Spoon, 21 cm
SDP
Catalog No. 5025718
Encompass_Preferred
This utensil's specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container. The Ellipso-Spoon Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Capacity: 21 cm
Size: 15 x 15 mm
Catalog No. 50-257-18 Supplier Electron Microscopy Sciences Supplier No. 7298421
This utensil's specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container. The Ellipso-Spoon Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Capacity: 21 cm
Size: 15 x 15 mm