Learn More
Chemglass Life Sciences Elliptical Bump Trap, Rotary Evaporator, with Drain Holes, 24/40 Top Outer Jt, 24/40 Lower Inner Jt
SDP
Catalog No. 501215242
Encompass_Preferred
Elliptical Bump Trap, Rotary Evaporator, with Drain Holes, 24/40 Top Outer Jt, 24/40 Lower Inner Jt. Similar to CG-1320-E, but with the addition of (2) 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Rotary evaporator bump trap is used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of “bumping”. Unique elliptical shape provides large trap volume while decreasing the overall length when compared to standard spherical designs. This allows the use of larger recovery flasks in a rotary evaporator. Traps have a 24/40 top outer joint and a 24/40 lower inner joint.
Catalog No. 50-121-5242 Supplier Chemglass Life Sciences Supplier No. CG1321E10
Elliptical Bump Trap, Rotary Evaporator, with Drain Holes, 24/40 Top Outer Jt, 24/40 Lower Inner Jt. Similar to CG-1320-E, but with the addition of (2) 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Rotary evaporator bump trap is used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of “bumping”. Unique elliptical shape provides large trap volume while decreasing the overall length when compared to standard spherical designs. This allows the use of larger recovery flasks in a rotary evaporator. Traps have a 24/40 top outer joint and a 24/40 lower inner joint.