Learn More
Enzo Life Sciences Ellipticine (50mg). CAS: 519-23-3
SDP
Supplier:  Enzo Life Sciences BMLGR3150050
Encompass_Preferred
A DNA-intercalating antitumor agent. Inhibits DNA topoisomerase II. Inhibits the phosphorylation of p53 in Lewis lung carcinoma cells and induces apoptosis. Inhibits cytochrome P-450-1A1. Alternative name: 5,11-Dimethyl-6H-pyrido[4,3-b]carbazole. Purity: ≥98% (TLC). Solubility: Soluble in DMSO, DMF or ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0967