Learn More
Medchemexpress LLC HY-15753A 5mg Medchemexpress, Ellipticine (hydrochloride) CAS:5081-48-1 Purity:>98%
SDP
Catalog No. 502081792
Encompass_Preferred
Medchemexpress, HY-15753A 5mg Ellipticine (hydrochloride) CAS:5081-48-1 Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1792 Supplier Medchemexpress LLC Supplier No. HY15753A5MG
Medchemexpress, HY-15753A 5mg Ellipticine (hydrochloride) CAS:5081-48-1 Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.