Learn More
Selleck Chemical LLC Ellipticine hydrochloride
SDP
Supplier:  Selleck Chemical LLC S67905MG
Encompass_Preferred
Ellipticine Hydrochloride (NSC 71795 PZE) is a potent inhibitor of DNA topoisomerase II and forms covalent DNA adducts mediated by its oxidation with cytochromes P450 (CYP) and peroxidases Ellipticine Hydrochloride is a natural product isolated from the Australian evergreen tree of the Apocynaceae family with antineoplastic activity
Catalog No. 50-000-02309