Learn More
Medchemexpress LLC Ellipticine hydrochloride | 5081-48-1 | MFCD00050600 | 99.3% | 282.77 g·mol⁻1 | C17H15ClN2 | 10 MG
SDP
Catalog No. 5000288471
Encompass_Preferred
Ellipticine hydrochloride is the hydrochloride salt of ellipticine, a polycyclic alkaloid used in research as a topoisomerase II inhibitor with antineoplastic activity. It is supplied as a light yellow solid with high reported purity for laboratory use only.
  • Inhibits DNA topoisomerase II activity.
  • High purity (99.34%).
  • Light yellow solid physical form.
  • Available in small research pack sizes, including 10 mg.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) and -20°C (1 month).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-88471 Supplier Medchemexpress LLC Supplier No. HY15753A10MG
Ellipticine hydrochloride is the hydrochloride salt of ellipticine, a polycyclic alkaloid used in research as a topoisomerase II inhibitor with antineoplastic activity. It is supplied as a light yellow solid with high reported purity for laboratory use only.
  • Inhibits DNA topoisomerase II activity.
  • High purity (99.34%).
  • Light yellow solid physical form.
  • Available in small research pack sizes, including 10 mg.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) and -20°C (1 month).
  • For research use only; not for human or veterinary use.