Learn More
Enzo Life Sciences Ellmans Reagent (5g). CAS: 69-78-3
SDP
Catalog No. 502009078
Encompass_Preferred
A sensitive sulfhydryl reagent. Alternative name: 5,5-Dithio-bis(2-nitrobenzoic acid), DTNB. Purity: ≥98% (Titration). Solubility: Soluble in 100% ethanol or dioxane. Long Term Storage: +4°C.
Catalog No. 50-200-9078 Supplier Enzo Life Sciences Supplier No. ALX400034G005
A sensitive sulfhydryl reagent. Alternative name: 5,5-Dithio-bis(2-nitrobenzoic acid), DTNB. Purity: ≥98% (Titration). Solubility: Soluble in 100% ethanol or dioxane. Long Term Storage: +4°C.