Learn More
MSC Ellwood Safety Metatarsal Guards, Aluminum Alloy, Spring Device, 1 Pair
Catalog No. 5000796580
Encompass
These metatarsal guards offer protection for the foot. Constructed from durable aluminum alloy, they feature a universal fit and a spring device for secure placement.
  • Metatarsal guard type
  • Made from aluminum alloy
  • Silver color
  • Supplied as one pair
Catalog No. 50-007-96580 Supplier MSC Supplier No. 06119945
These metatarsal guards offer protection for the foot. Constructed from durable aluminum alloy, they feature a universal fit and a spring device for secure placement.
  • Metatarsal guard type
  • Made from aluminum alloy
  • Silver color
  • Supplied as one pair