Learn More
Enterprise Technology Solutions Elmer's Self Healing Cutting Mats, 12"x18", Opaque Gray
SDP
Catalog No. 5000831394
Encompass_Preferred
Heavy-duty, three-ply Self-healing Cutting Mat features a special surface and core that work together to minimize effects of cutting with sharp blades. The 1" grid pattern with 90 degree, 45 degree and 30 degree angular guides ensures an accurate layout. Eighths are clearly printed along edge guides. Ideal for art, drafting and engineering students or anyone with a cutting project.
Catalog No. 50-008-31394 Supplier Enterprise Technology Solutions Supplier No. EPIX7761
Heavy-duty, three-ply Self-healing Cutting Mat features a special surface and core that work together to minimize effects of cutting with sharp blades. The 1" grid pattern with 90 degree, 45 degree and 30 degree angular guides ensures an accurate layout. Eighths are clearly printed along edge guides. Ideal for art, drafting and engineering students or anyone with a cutting project.