Learn More
Electron Microscopy Sciences Rubber Cement, 4oz with brush in cap
SDP
Catalog No. 501899750
Encompass_Preferred
Excellent adhesive for all paper pasting and mountings of drawings and photographs. It is also used to adhere specimens to SEM Stubs. Any excess cured cement can be easily removed. 4 oz bottle comes with a brush in the cap.
Catalog No. 50-189-9750 Supplier Electron Microscopy Sciences Supplier No. 72170
Excellent adhesive for all paper pasting and mountings of drawings and photographs. It is also used to adhere specimens to SEM Stubs. Any excess cured cement can be easily removed. 4 oz bottle comes with a brush in the cap.