Learn More
Cayman Chemical Ellman's Reagent, 100 dtn
SDP
Catalog No. 502945925
Encompass_Preferred
A substrate for all AChE-based ELISA kits.
Catalog No. 50-294-5925 Supplier Cayman Chemical Supplier No. 400050100 DTN
A substrate for all AChE-based ELISA kits.