Learn More
Aviva Systems Biology Corp ELMO1 Antibody - C-terminal region (ARP59647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELMO1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ELMO1. Alternate names: CED-12, CED12, ELMO-1, KIAA0281, MGC126406ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-766