Learn More
Novus Biologicals™ ELMO2 Peptide

Specifications
Specifications
| Gene ID (Entrez) | 63916 |
| Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-879 |
| Gene Alias | ced-12 homolog 2, CED12A, ELMO-2, engulfment and cell motility 2, engulfment and cell motility 2 (ced-12 homolog, C. elegans), engulfment and cell motility protein 2, FLJ11656, hCed-12 A, KIAA1834CED12CED-12, PH domain protein CED12A, Protein ced-12 homolog A |
| Gene Symbol | ELMO2 |
| Quantity | 0.1 mg |
| Research Category | Apoptosis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.