Learn More
Aviva Systems Biology Corp ELMO3 Antibody - middle region (ARP42635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELMO3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ELMO3. Alternate names: CED-12, CED12, ELMO-3, FLJ13824ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58287