Learn More
Aviva Systems Biology Corp ELMOD3 Antibody - C-terminal region (ARP60431_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60431_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELMOD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ELMOD3. Alternate names: FLJ21977, FLJ35601, LST3, MGC111036, RBED1, RBM29ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-040