Learn More
Medchemexpress LLC Eln318463 | 851600-86-7 | 99.5% | 471.8 g/mol | C19H20BrClN2O3S | 10 MG
SDP
Catalog No. 5000297957
Encompass_Preferred
This compound is an amyloid precursor protein (APP)-selective γ-secretase inhibitor for in vitro research. It preferentially inhibits PS1-containing γ-secretase (EC50 = 12 nM) over PS2 (EC50 = 656 nM), providing ~51-fold selectivity. The solid is high purity, soluble in DMSO, and intended for biochemical and cellular assay applications.
  • High purity (99.5%).
  • APP-selective γ-secretase inhibitory activity.
  • Potent PS1 inhibition (EC50 12 nM), reduced PS2 activity.
  • Soluble in DMSO for assay preparation.
  • Suitable for biochemical and cellular assays.
Catalog No. 50-002-97957 Supplier Medchemexpress LLC Supplier No. HY5088210MG
This compound is an amyloid precursor protein (APP)-selective γ-secretase inhibitor for in vitro research. It preferentially inhibits PS1-containing γ-secretase (EC50 = 12 nM) over PS2 (EC50 = 656 nM), providing ~51-fold selectivity. The solid is high purity, soluble in DMSO, and intended for biochemical and cellular assay applications.
  • High purity (99.5%).
  • APP-selective γ-secretase inhibitory activity.
  • Potent PS1 inhibition (EC50 12 nM), reduced PS2 activity.
  • Soluble in DMSO for assay preparation.
  • Suitable for biochemical and cellular assays.