Learn More
Medchemexpress LLC HY-115038 5mg Medchemexpress, ELN484228 CAS:312-63-0 Purity:>98%
SDP
Catalog No. 501969377
Encompass_Preferred
Medchemexpress, HY-115038 5mg ELN484228 CAS:312-63-0 ELN484228 is a blocker of α-synuclein which is a key protein in Parkinson’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9377 Supplier Medchemexpress LLC Supplier No. HY1150385MG
Medchemexpress, HY-115038 5mg ELN484228 CAS:312-63-0 ELN484228 is a blocker of α-synuclein which is a key protein in Parkinson’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.