Learn More
Aviva Systems Biology Corp ELOF1 Antibody - N-terminal region (ARP60432_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60432_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELOF1. It was validated on Western Blot. . Alternate names: ELF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-041