Learn More
Selleck Chemical LLC Elotuzumab-A2034-1MG
SDP
Supplier:  Selleck Chemical LLC A20341MG
Encompass_Preferred
Elotuzumab (anti-SLAMF7 BMS-901608 PDL063 HuLuc63) is a humanized monoclonal antibody that binds human signaling lymphocytic activation molecule F7 (hSLAMF7 also known as CS1 CD319 or CRACC) Elotuzumab shows a dual mechanism of action that includes the direct activation of natural killer (NK) cells and the induction of NK cell mediated antibody-dependent cellular cytotoxicity
Catalog No. 50-234-0944