Learn More
TARGETMOL CHEMICALS INC ELOVL1-IN-2 5MG
SDP
Catalog No. 502579599
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ELOVL1-IN-2 a compound known as an inhibitor of the elongation of very long chain fatty acid 1 (ELOVL1) enzyme exhibits a weak inhibitory effect on ELOVL1 (with an IC50 of 21 uM) and demonstrates moderate potency in a primary cellular assay conducted on HEK293 C26 cells (with an IC50 of 6.7 uM) [1]. purity: 99%
Catalog No. 50-257-9599 Supplier TARGETMOL CHEMICALS INC Supplier No. T606275MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ELOVL1-IN-2 a compound known as an inhibitor of the elongation of very long chain fatty acid 1 (ELOVL1) enzyme exhibits a weak inhibitory effect on ELOVL1 (with an IC50 of 21 uM) and demonstrates moderate potency in a primary cellular assay conducted on HEK293 C26 cells (with an IC50 of 6.7 uM) [1]. purity: 99%