Learn More
Medchemexpress LLC HY-145272 1mg , ELOVL1-IN-3 CAS:2761063-99-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1452721MG
Encompass_Preferred
HY-145272 1mg ELOVL1-IN-3 CAS: 2761063-99-2 ELOVL1-IN-3 (Compound 22) is a potent and orally active inhibitor of elongation of very long chain fatty acid 1 (ELOVL1) enzyme. ELOVL1-IN-3 serves as a useful tool for researching adrenoleukodystrophy (ALD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1856