Learn More
Aviva Systems Biology Corp ELOVL6 Antibody - N-terminal region (ARP64566_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64566_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELOVL6. It was validated on Western Blot. . Alternate names: FACE, FAE, FLJ23378, LCE, MGC5487ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-300