Learn More
Aviva Systems Biology Corp ELOVL7 Antibody - N-terminal region (ARP49908_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49908_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ELOVL7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ELOVL7. Alternate names: FLJ23563ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53608