Learn More
Aviva Systems Biology Corp ELP2 Antibody - middle region (ARP57179_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57179_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ELP2. Alternate names: FLJ10879, SHINC-2, STATIP1, StIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53958