Learn More
ABclonal Technology ELP2 Rabbit pAb
SDP
Catalog No. 5000106655
Encompass_Preferred
The protein encoded by this gene is a core subunit of the elongator complex, a histone acetyltransferase complex that associates with RNA polymerase II. In addition to histone acetylation, the encoded protein effects transcriptional elongation and may help remodel chromatin.
Catalog No. 50-001-06655 Supplier ABclonal Technology Supplier No. A15857500UL
The protein encoded by this gene is a core subunit of the elongator complex, a histone acetyltransferase complex that associates with RNA polymerase II. In addition to histone acetylation, the encoded protein effects transcriptional elongation and may help remodel chromatin.