Learn More
Corning™ Elplasia™ 96-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
GREENER_CHOICE
Catalog No. 07202463 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-202-463 Black, Clear Bottom
1 options
This item is not returnable. View return policy
Catalog No. 07-202-463 Supplier Corning™ Supplier No. 4442
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
The only plates with a simple “plug and play” protocol for scaffold-free, self-assembly spheroid formation at large volumes.
Novel well geometry and surface coatings, including Corning Ultra Low Attachment (ULA) featured on the round bottom plates. Corning ULA surface is hydrophilic, biologically inert and non-degradable, which promotes highly reproducible spheroid formation and easy harvesting.
Corning Elplasia round bottom plates optimal for bulk spheroid formation, collection and expansion.  Round bottom plates feature Corning Ultra-Low Attachment surface and are available in 6-, 24- and 96-well formats.
•Spheroids may be formed and cultured for 21 or more days
•Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
•The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
•Black opaque sidewalls to reduce well-to-well “cross-talk”
•Well suited for fluorescent/luminescent assays
​•Available in multiple formats, two well geometries and surface coating options.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Breakapart No
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Ultra-Low Attachment
Color Black, Clear Bottom
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round Bottom (U-well)
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5
Skirt Color Black
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov