Learn More
Selleck Chemical LLC ELR-510444 S3725-5mg
SDP
Catalog No. 503108378
Encompass_Preferred
ELR-510444 is a novel microtubule disruptor with potential antivascular effects and in vivo antitumor efficacy causing a loss of cellular microtubules and the formation of aberrant mitotic spindles and leading to mitotic arrest and apoptosis of cancer cells
Catalog No. 50-310-8378 Supplier Selleck Chemical LLC Supplier No. S37255MG
ELR-510444 is a novel microtubule disruptor with potential antivascular effects and in vivo antitumor efficacy causing a loss of cellular microtubules and the formation of aberrant mitotic spindles and leading to mitotic arrest and apoptosis of cancer cells