Learn More
Selleck Chemical LLC Elraglusib S9602-25MG
SDP
Catalog No. 5000024073
Encompass_Preferred
Elraglusib(9-ING-41) is a potent inhibitor of glycogen synthase kinase-3 (GSK-3) with antitumor activity 9-ING-41 induces apoptosis and cell cycle arrest at prophase by targeting centrosomes and microtubule-bound GSK3
Catalog No. 50-000-24073 Supplier Selleck Chemical LLC Supplier No. S960225MG
Elraglusib(9-ING-41) is a potent inhibitor of glycogen synthase kinase-3 (GSK-3) with antitumor activity 9-ING-41 induces apoptosis and cell cycle arrest at prophase by targeting centrosomes and microtubule-bound GSK3