Learn More
3M™ Scott™ ELSA™ Emergency Escape Breathing Device - N (EEBD-N)
Catalog No. p-7222315
Change view
Click to view available options
Certifications/Compliance:
NIOSH
NIOSH, SOLAS Chapter II-2 Part D
Includes:
Respirator, Hood, Carry Bag, Drained Cylinder
Respirator, Hood, Carry Bag, Full Cylinder
Duration:
10 min.
15 min.
5 min.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Includes Duration
19-072-397 NIOSH Respirator, Hood, Carry Bag, Full Cylinder 5 min.
19-072-398 NIOSH, SOLAS Chapter II-2 Part D Respirator, Hood, Carry Bag, Full Cylinder 10 min.
19-072-401 NIOSH, SOLAS Chapter II-2 Part D Respirator, Hood, Carry Bag, Drained Cylinder 10 min.
19-072-399 NIOSH, SOLAS Chapter II-2 Part D Respirator, Hood, Carry Bag, Full Cylinder 15 min.
19-072-402 NIOSH, SOLAS Chapter II-2 Part D Respirator, Hood, Carry Bag, Drained Cylinder 15 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-072-397 Supplier 3M™ Scott™ Supplier No. 7012473800
Please contact your local Fisher Scientific sales representative for pricing
ELSA™ EEBD-N respirator emergency escape breathing device is designed for simple use, training, and maintenance. Carry bag offers improved functionality while the redesigned hood fits wearers comfortably.
Enables workers to escape from certain toxic environments, even in atmospheres immediately dangerous to life or health (IDLH).
  • Features one-size-fits-all clear hood with patent-pending neck seal
  • Pulls on with no additional steps to secure, making it easy to train and use properly
  • Includes fully-enclosed, chemical-resistant PVC carry bag to keep it free of insects and debris
  • Available with 5, 10, or 15 minute durations
  • Available with full or drained cylinders
  • NIOSH approved
  • 10 and 15 minute models meet SOLAS Chapter II-2, Part D requirements.
  • Fits into standard wall cases, has indefinite shelf life, and comes with a 3 year warranty. 
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Product Type Escape Breathing Apparatus
Duration 5 min.
Pressure 30 lpm
Hood Type Clear hood with patent-pending neck seal fits all wearers, and pulls on with no additional steps to secure, making it easy to train and use properly.
Includes Respirator, Hood, Carry Bag, Full Cylinder
Model 8007212