Learn More
Waters Corp Waters ELS Detector Standard, Liquid, 10 mL, Acetaminophen in Methanol
Catalog No. 50792745 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ELS Detector Standard is a liquid solution for calibrating and validating Evaporative Light Scattering Detectors (ELSDs) in HPLC. It combines acetaminophen and methanol, offering a universal response for compounds and overcoming UV or fluorescence detection limits.
  • Calibrates and validates ELSDs in HPLC applications.
  • Contains acetaminophen in methanol for precise calibration.
  • Provides universal compound detection, surpassing UV/fluorescence limits.
  • 10 mL ampule for convenient use.
  • Compatible with isocratic and gradient separations.
  • Verifies ongoing ELSD performance.
  • Streamlines calibration and validation, optimizing workflows.
  • Cost-effective, accurate solution with a 24-month shelf life.
  • Stores at room temperature.
Catalog No. 50-792-745 Supplier Waters Corp Supplier No. 700002387
The ELS Detector Standard is a liquid solution for calibrating and validating Evaporative Light Scattering Detectors (ELSDs) in HPLC. It combines acetaminophen and methanol, offering a universal response for compounds and overcoming UV or fluorescence detection limits.
  • Calibrates and validates ELSDs in HPLC applications.
  • Contains acetaminophen in methanol for precise calibration.
  • Provides universal compound detection, surpassing UV/fluorescence limits.
  • 10 mL ampule for convenient use.
  • Compatible with isocratic and gradient separations.
  • Verifies ongoing ELSD performance.
  • Streamlines calibration and validation, optimizing workflows.
  • Cost-effective, accurate solution with a 24-month shelf life.
  • Stores at room temperature.