Learn More
TARGETMOL CHEMICALS INC ELSIBUCOL 5MG
SDP
Catalog No. 502580359
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Elsibucol (AGI 1096) is a VCAM1 inhibitor for the study of organ transplant rejection.Elsibucol is a metabolically stable propanol derivative with antioxidant anti-inflammatory and anti-proliferative properties. It lowers blood cholesterol levels and reduces oxidative stress and inflammatory responses in injured arteries thereby inhibiting atherosclerosis and protecting endothelial healing after arterial injury. purity: 98%
Catalog No. 50-258-0359 Supplier TARGETMOL CHEMICALS INC Supplier No. T316155MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Elsibucol (AGI 1096) is a VCAM1 inhibitor for the study of organ transplant rejection.Elsibucol is a metabolically stable propanol derivative with antioxidant anti-inflammatory and anti-proliferative properties. It lowers blood cholesterol levels and reduces oxidative stress and inflammatory responses in injured arteries thereby inhibiting atherosclerosis and protecting endothelial healing after arterial injury. purity: 98%