Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Elsulfavirine | 868046-19-9 | MFCD32215300 | 99.9% | 629.28 g·mol⁻¹ | C24H17BrCl2FN3O5S | 5 MG
SDP

Catalog No. 5000264625
Encompass_Preferred

Elsulfavirine is a non-nucleoside reverse transcriptase inhibitor (NNRTI) investigated for treatment of HIV-1. This listing is a research-grade reference compound supplied as a small solid amount for laboratory in vitro and in vivo studies; it is not intended for clinical use.

  • Non-nucleoside reverse transcriptase inhibitor activity.
  • Research-grade compound for laboratory use.
  • High reported purity: 99.9%.
  • Supplied as a 5 mg solid.
  • Molecular weight approximately 629.28 g·mol⁻¹; formula C24H17BrCl2FN3O5S.
  • Powder storage: -20°C; solution storage guidance available.
  • Soluble in DMSO (~62.5 mg/mL); may require sonication.
  • Not for human or veterinary use.

Catalog No. 50-002-64625 Supplier Medchemexpress LLC Supplier No. HY1090565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Elsulfavirine is a non-nucleoside reverse transcriptase inhibitor (NNRTI) investigated for treatment of HIV-1. This listing is a research-grade reference compound supplied as a small solid amount for laboratory in vitro and in vivo studies; it is not intended for clinical use.

  • Non-nucleoside reverse transcriptase inhibitor activity.
  • Research-grade compound for laboratory use.
  • High reported purity: 99.9%.
  • Supplied as a 5 mg solid.
  • Molecular weight approximately 629.28 g·mol⁻¹; formula C24H17BrCl2FN3O5S.
  • Powder storage: -20°C; solution storage guidance available.
  • Soluble in DMSO (~62.5 mg/mL); may require sonication.
  • Not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.