Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eltanexor Z-isomer | 1642300-78-4 | 96.0% | 428.29 g/mol | C17H10F6N6O | 1 ML
SDP

Catalog No. 5000214003
Encompass_Preferred

Eltanexor Z-isomer is the less-active geometric isomer of eltanexor, provided for research use as a ready-to-use 10 mM solution in DMSO. It is intended for laboratory studies of exportin-1 (XPO1/CRM1) inhibition, biochemical assays, and reference analyses.

  • Provided as 10 mM solution in DMSO, 1 mL.
  • Purity approximately 96.0% suitable for research applications.
  • Molecular weight 428.29 g/mol and formula C17H10F6N6O.
  • CAS number 1642300-78-4 for substance identification.
  • Appearance light yellow solid; solution supplied in DMSO.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Catalog No. 50-002-14003 Supplier Medchemexpress LLC Supplier No. HY100423A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eltanexor Z-isomer is the less-active geometric isomer of eltanexor, provided for research use as a ready-to-use 10 mM solution in DMSO. It is intended for laboratory studies of exportin-1 (XPO1/CRM1) inhibition, biochemical assays, and reference analyses.

  • Provided as 10 mM solution in DMSO, 1 mL.
  • Purity approximately 96.0% suitable for research applications.
  • Molecular weight 428.29 g/mol and formula C17H10F6N6O.
  • CAS number 1642300-78-4 for substance identification.
  • Appearance light yellow solid; solution supplied in DMSO.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.