Learn More
Medchemexpress LLC Eltanexor Z-isomer | 1642300-78-4 | 96.0% | 428.29 g/mol | C17H10F6N6O | 100 MG
SDP
Catalog No. 5000213939
Encompass_Preferred
Eltanexor Z-isomer is the geometric (Z) isomer of eltanexor, a small-molecule exportin-1 (XPO1/CRM1) inhibitor. Supplied for laboratory research use only, the compound is offered as a solid and as DMSO solutions across multiple pack sizes and is intended for in vitro studies and research applications.
  • Less active geometric isomer of the parent compound.
  • High purity suitable for research applications.
  • Available as solid and as DMSO solution for convenience.
  • Characterized by molecular formula and molecular weight for traceability.
  • Intended for laboratory research use only; not for human or clinical use.
Catalog No. 50-002-13939 Supplier Medchemexpress LLC Supplier No. HY100423A100MG
Eltanexor Z-isomer is the geometric (Z) isomer of eltanexor, a small-molecule exportin-1 (XPO1/CRM1) inhibitor. Supplied for laboratory research use only, the compound is offered as a solid and as DMSO solutions across multiple pack sizes and is intended for in vitro studies and research applications.
  • Less active geometric isomer of the parent compound.
  • High purity suitable for research applications.
  • Available as solid and as DMSO solution for convenience.
  • Characterized by molecular formula and molecular weight for traceability.
  • Intended for laboratory research use only; not for human or clinical use.