Learn More
Medchemexpress LLC Eltoprazine hydrochloride | 98206-09-8 | 99.3% | 256.73 | 100 MG
SDP
Catalog No. 5000393703
Encompass_Preferred
Eltoprazine hydrochloride is an agonist for 5-HT1A/5-HT1B receptors and an antagonist for 5-HT2C receptors, demonstrating antiaggressive and anxiogenic effects.
  • Agonist for 5-HT1A/5-HT1B receptors
  • Antagonist for 5-HT2C receptors
  • Shows antiaggressive effects
  • Shows anxiogenic effects
  • Target: 5-HT Receptor
  • Pathway: GPCR/G Protein, Neuronal Signaling
Catalog No. 50-003-93703 Supplier Medchemexpress LLC Supplier No. HY16687A100MG
Eltoprazine hydrochloride is an agonist for 5-HT1A/5-HT1B receptors and an antagonist for 5-HT2C receptors, demonstrating antiaggressive and anxiogenic effects.
  • Agonist for 5-HT1A/5-HT1B receptors
  • Antagonist for 5-HT2C receptors
  • Shows antiaggressive effects
  • Shows anxiogenic effects
  • Target: 5-HT Receptor
  • Pathway: GPCR/G Protein, Neuronal Signaling