Learn More
Medchemexpress LLC Eltoprazine hydrochloride | 98206-09-8 | 99.3% | 256.73 | 5 MG
SDP
Catalog No. 5000393814
Encompass_Preferred
Eltoprazine hydrochloride (DU 28853) is a 5-HT1A/5-HT1B receptors agonist and a 5-HT2C receptor antagonist. It shows antiaggressive and anxiogenic effects.
  • Acts as a 5-HT1A/5-HT1B receptors agonist.
  • Acts as a 5-HT2C receptor antagonist.
  • Shows antiaggressive and anxiogenic effects.
  • For research use only.
Catalog No. 50-003-93814 Supplier Medchemexpress LLC Supplier No. HY16687A5MG
Eltoprazine hydrochloride (DU 28853) is a 5-HT1A/5-HT1B receptors agonist and a 5-HT2C receptor antagonist. It shows antiaggressive and anxiogenic effects.
  • Acts as a 5-HT1A/5-HT1B receptors agonist.
  • Acts as a 5-HT2C receptor antagonist.
  • Shows antiaggressive and anxiogenic effects.
  • For research use only.