Learn More
Medchemexpress LLC Eltoprazine hydrochloride | 98206-09-8 | 99.3% | 256.73 | 50 MG
Supplier: Medchemexpress LLC HY16687A50MG
Eltoprazine (DU 28853) hydrochloride is an agonist for 5-HT1A/5-HT1B receptors and an antagonist for 5-HT2C receptors. It has demonstrated antiaggressive and anxiogenic effects.
- Agonist for 5-HT1A/5-HT1B receptors.
- Antagonist for 5-HT2C receptors.
- Shows antiaggressive and anxiogenic effects.
- High purity of 99.25%.