Learn More
Cayman Chemical EltoprazInhydrochlorIde 10mg
SDP
Catalog No. 502967672
Encompass_Preferred
A partial agonist at 5-HT1A, 5-HT1B, and 5-HT2B receptors (Kis = 40, 52, and 81 nM, respectively); decreases aggressive behavior in resident-intruder tests with rats (ID50 = 0.24 mg/kg)
Catalog No. 50-296-7672 Supplier Cayman Chemical Supplier No. 1842810 MG
A partial agonist at 5-HT1A, 5-HT1B, and 5-HT2B receptors (Kis = 40, 52, and 81 nM, respectively); decreases aggressive behavior in resident-intruder tests with rats (ID50 = 0.24 mg/kg)