Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eltrombopag olamine | 496775-61-2 | ≥98.0% | 442.47 g/mol | C25H22N4O4 | 10 MG
SDP

Catalog No. 502029114
Encompass_Preferred

Eltrombopag is an orally active thrombopoietin (TPO) receptor nonpeptide agonist used in research to stimulate megakaryopoiesis and study thrombocytopenia, hematology, and oncology-related mechanisms. Supplied as a research-grade compound for use in in vitro and in vivo studies.

  • Stimulates thrombopoietin receptor signaling to increase platelet production.
  • Suitable for in vitro and in vivo research applications.
  • Available as the olamine salt for improved handling.
  • Typical purity ≥98.0% by HPLC as reported by suppliers.
  • Molecular formula C25H22N4O4, molecular weight 442.47 g/mol.
  • CAS number 496775-61-2 for the parent compound.

Catalog No. 50-202-9114 Supplier Medchemexpress LLC Supplier No. HY1530610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eltrombopag is an orally active thrombopoietin (TPO) receptor nonpeptide agonist used in research to stimulate megakaryopoiesis and study thrombocytopenia, hematology, and oncology-related mechanisms. Supplied as a research-grade compound for use in in vitro and in vivo studies.

  • Stimulates thrombopoietin receptor signaling to increase platelet production.
  • Suitable for in vitro and in vivo research applications.
  • Available as the olamine salt for improved handling.
  • Typical purity ≥98.0% by HPLC as reported by suppliers.
  • Molecular formula C25H22N4O4, molecular weight 442.47 g/mol.
  • CAS number 496775-61-2 for the parent compound.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.