Learn More
Cayman Chemical Eltrombopag 10mg
SDP
Catalog No. 502952777
Encompass_Preferred
A thrombopoietin receptor agonist (EC50 = 0.27 µM) and iron chelator; It increases STAT5 phosphorylation in N2C-Tpo cells at 30 µM and p42/44 MAPK phosphorylation at 10 µM; stimulates megakaryocytopoiesis in human primary bone marrow cells; increases platelet production in chimpanzees at 10 mg/kg per day for five days; binds to iron (III) and mobilizes cellular iron and ferritin in H9C2, Huh7, and RINm5F cells
Catalog No. 50-295-2777 Supplier Cayman Chemical Supplier No. 1324710 MG
A thrombopoietin receptor agonist (EC50 = 0.27 µM) and iron chelator; It increases STAT5 phosphorylation in N2C-Tpo cells at 30 µM and p42/44 MAPK phosphorylation at 10 µM; stimulates megakaryocytopoiesis in human primary bone marrow cells; increases platelet production in chimpanzees at 10 mg/kg per day for five days; binds to iron (III) and mobilizes cellular iron and ferritin in H9C2, Huh7, and RINm5F cells