Learn More
Medchemexpress LLC Eltrombopag-13C4 | 1217230-31-3 | C2113C4H22N4O4 | 1 MG
SDP
Catalog No. 5000374176
Encompass_Preferred
Eltrombopag-13C4 (SB-497115-13C4) is 13C-labeled Z-Eltrombopag. Z-Eltrombopag is an orally active thrombopoietin-receptor non-peptide agonist with platelet-stimulating activity for the study of chronic immune thrombocytopenia. Eltrombopag also has strong inhibitory effects on multidrug-resistant Staphylococcus aureus and can induce apoptosis in liver cancer cells.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Acts as an orally active thrombopoietin-receptor non-peptide agonist.
  • Exhibits platelet-stimulating activity, useful for studying chronic immune thrombocytopenia.
  • Demonstrates strong inhibitory effects on multidrug-resistant Staphylococcus aureus.
  • Induces apoptosis in liver cancer cells.
Catalog No. 50-003-74176 Supplier Medchemexpress LLC Supplier No. HY15306S1MG
Eltrombopag-13C4 (SB-497115-13C4) is 13C-labeled Z-Eltrombopag. Z-Eltrombopag is an orally active thrombopoietin-receptor non-peptide agonist with platelet-stimulating activity for the study of chronic immune thrombocytopenia. Eltrombopag also has strong inhibitory effects on multidrug-resistant Staphylococcus aureus and can induce apoptosis in liver cancer cells.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Acts as an orally active thrombopoietin-receptor non-peptide agonist.
  • Exhibits platelet-stimulating activity, useful for studying chronic immune thrombocytopenia.
  • Demonstrates strong inhibitory effects on multidrug-resistant Staphylococcus aureus.
  • Induces apoptosis in liver cancer cells.